Assay Details
| Gene Symbol: Rbpj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106631.1 | NP_001100101.1 | 3 - 5 | 215 | 76 | |
| XM_008770190.1 | XP_008768412.1 | 2 - 4 | 886 | 76 | ||
| XM_008770191.1 | XP_008768413.1 | 2 - 4 | 127 | 76 | ||
| XM_008770192.1 | XP_008768414.1 | 2 - 4 | 2305 | 76 | ||
| XM_008770193.1 | XP_008768415.1 | 3 - 5 | 298 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106631.1 transcripts.
Set Membership: |
|

Rbpj
Genomic Map