This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01457054_m1
See other PMP2 GE Assays ›
Gene Symbol:
Pmp2
Gene Name:
peripheral myelin protein 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 93827504 - 93831419 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01457054_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pmp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109514.1 NP_001102984.1 1 - 2 121 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200000780.rn.1

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transport
membrane organization
transporter activity
fatty acid binding
cholesterol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200004826.rn.1 TC0200000780.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200004827.rn.1 TC0200000780.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200004828.rn.1 TC0200000780.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline