This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01457229_g1
See other GTF3A GE Assays ›
Gene Symbol:
Gtf3a
Gene Name:
general transcription factor III A
Gene Aliases:
-
Chromosome Location:
Chr.12: 9856841 - 9864791 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01457229_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113570.1 NP_001107042.1 7 - 8 1025 65
GenBank mRNA CB578423.1 - 2 - 3 65

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
RNA binding
metal ion binding

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P262025 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P262025 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P262025 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P262025 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P262025 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1395970_at - -
Rat Genome 230 2.0 Array 1371035_PM_at - -
Rat Genome 230 2.0 Array 1395970_PM_at - -
Rat Genome 230 2.0 Array 1371035_at - -
Affymetrix HT Rat Focus Array 1371035_at - -
Affymetrix HT Rat Focus Array 1371035_PM_at - -
Affymetrix Rat Clariom S Assay PSR1200008884.rn.1 TC1200001054.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200008885.rn.1 TC1200001054.rn.1 RTA TC
Rat Expression Set 230 1371035_at - -
Rat Expression Set 230 1371035_PM_at - -
Rat Genome U34 Set rc_AI170270_at - -
Rat Genome U34 Set rc_AI236565_at - -
Rat Genome U34 Set rc_AI178189_s_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22297123 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline