Assay Details
| Gene Symbol: Psg29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025641.1 | NP_001020812.1 | 1 - 2 | 188 | 91 | |
| GenBank mRNA | BC097354.1 | - | 1 - 2 | 188 | 7441714 | 91 |
More Information
Updated Mapping Information
Set Membership: |
|

Psg29
Genomic Map