This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01458747_g1
See other PPID GE Assays ›
Gene Symbol:
Ppid
Gene Name:
peptidylprolyl isomerase D
Gene Aliases:
Cyp-40, CypD
Chromosome Location:
Chr.2: 178354830 - 178366843 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01458747_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppid
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004279.1 NP_001004279.1 6 - 7 817 198
GenBank mRNA BC076386.1 - 6 - 7 817 7108860 198
FQ214826.1 - 6 - 7 820 198

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppid

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
protein peptidyl-prolyl isomerization
protein folding
protein complex assembly
apoptotic process
response to oxidative stress
protein transport
lipid particle organization
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of viral genome replication
positive regulation of protein secretion
regulation of mitochondrial membrane potential
chaperone-mediated protein folding
cellular response to UV-A
peptidyl-prolyl cis-trans isomerase activity
protein binding
transcription factor binding
cyclosporin A binding
enzyme binding
estrogen receptor binding
Hsp70 protein binding
heat shock protein binding
Hsp90 protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200008727.rn.1 TC0200001429.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200008729.rn.1 TC0200001429.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline