This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01459205_m1
See other GPR162 GE Assays ›
Gene Symbol:
Gpr162
Gene Name:
G protein-coupled receptor 162
Gene Aliases:
Grca
Chromosome Location:
Chr.4: 157375184 - 157381779 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01459205_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr162
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108646.1 NP_001102116.1 3 - 4 1735 62
XM_006237371.2 XP_006237433.1 3 - 4 1832 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0400003742.rn.1

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P246081 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 873160 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 224011 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 627044 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 234593 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 558458 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 124373 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P246081 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P246081 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P246081 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P246081 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10865579 - -
Affymetrix Rat Gene 1.0 ST Array 10865581 - -
Affymetrix Rat Gene 1.0 ST Array 10865584 - -
Affymetrix Rat Gene 1.0 ST Array 10865582 - -
Affymetrix Rat Gene 1.0 ST Array 10865583 - -
Affymetrix Rat Gene 1.0 ST Array 10865580 - -
Affymetrix Rat Gene 1.1 ST Array 10865578 - -
Rat Genome 230 2.0 Array 1374445_PM_at - -
Rat Genome 230 2.0 Array 1374445_at - -
Affymetrix HT Rat Focus Array 1374445_at - -
Affymetrix HT Rat Focus Array 1374445_PM_at - -
Affymetrix Rat Clariom S Assay PSR0400026574.rn.1 TC0400003742.rn.1 RTA TC
Rat Expression Set 230 1374445_at - -
Rat Expression Set 230 1374445_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10865578 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21336182 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1356547 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0