This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01462700_m1
See other FGF23 GE Assays ›
Gene Symbol:
Fgf23
Gene Name:
fibroblast growth factor 23
Gene Aliases:
-
Chromosome Location:
Chr.4: 159622404 - 159630082 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01462700_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer:
284.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fgf23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130754.1 NP_570110.1 2 - 3 317 82
GenBank mRNA AB078777.1 - 2 - 3 317 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0400001814.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fgf23

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

phosphate-containing compound metabolic process
fibroblast growth factor receptor signaling pathway
regulation of phosphate transport
positive regulation of vitamin D 24-hydroxylase activity
cell differentiation
regulation of bone mineralization
negative regulation of bone mineralization
cellular phosphate ion homeostasis
vitamin D metabolic process
vitamin D catabolic process
negative regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
negative regulation of hormone secretion
phosphate ion homeostasis
positive regulation of ERK1 and ERK2 cascade
positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
fibroblast growth factor receptor binding
type 1 fibroblast growth factor receptor binding
growth factor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P146675 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 609509 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 323739 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 767956 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 586214 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 713648 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 386341 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 91016 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P146675 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P146675 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P146675 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P146675 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 77873 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10859005 - -
Affymetrix Rat Gene 1.0 ST Array 10859006 - -
Affymetrix Rat Gene 1.0 ST Array 10859004 - -
Affymetrix Rat Gene 1.1 ST Array 10859003 - -
Rat Genome 230 2.0 Array 1369595_at - -
Rat Genome 230 2.0 Array 1369595_PM_at - -
Affymetrix HT Rat Focus Array 1369595_at - -
Affymetrix HT Rat Focus Array 1369595_PM_at - -
Affymetrix Rat Clariom S Assay PSR0400013346.rn.1 TC0400001814.rn.1 RTA TC
Rat Expression Set 230 1369595_PM_at - -
Rat Expression Set 230 1369595_at - -
Affymetrix Rat Gene 1.0 ST Array 10859003 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22422551 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1366217 - -
RatRef-12 Expression BeadChip ILMN_60868 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline