This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01464076_m1
See other TOM1L2 GE Assays ›
Gene Symbol:
Tom1l2
Gene Name:
target of myb1-like 2 (chicken)
Gene Aliases:
-
Chromosome Location:
Chr.10: 46599392 - 46720913 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01464076_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tom1l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108277.1 NP_001101747.1 4 - 5 510 62
XM_006246506.2 XP_006246568.1 5 - 6 592 62
XM_006246507.2 XP_006246569.1 5 - 6 592 62
XM_006246508.2 XP_006246570.1 5 - 6 592 62
XM_008767833.1 XP_008766055.1 5 - 6 592 62
XM_008767834.1 XP_008766056.1 5 - 6 592 62
XM_008767835.1 XP_008766057.1 5 - 6 2125 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000002931.rn.1

Set Membership:

Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
signal transduction
negative regulation of mitotic nuclear division
protein kinase binding
clathrin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000025755.rn.1 TC1000002931.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000025759.rn.1 TC1000002931.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000025756.rn.1 TC1000002931.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000025754.rn.1 TC1000002931.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000025757.rn.1 TC1000002931.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000025764.rn.1 TC1000002931.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0