This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01464129_g1
See other PRSS21 GE Assays ›
Gene Symbol:
Prss21
Gene Name:
protease, serine, 21
Gene Aliases:
Esp-1, Resp-1
Chromosome Location:
Chr.10: 13214395 - 13220052 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01464129_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss21
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181477.2 NP_852142.1 5 - 6 763 72
GenBank mRNA AB074516.1 - 5 - 6 763 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000000192.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prss21

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
spermatogenesis
serine-type endopeptidase activity
serine-type peptidase activity

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1052006 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1052006 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1052006 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1052006 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1052006 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10940587 - -
Affymetrix Rat Gene 1.1 ST Array 10940587 - -
Rat Genome 230 2.0 Array 1375328_PM_at - -
Rat Genome 230 2.0 Array 1375328_at - -
Affymetrix HT Rat Focus Array 1375328_PM_at - -
Affymetrix HT Rat Focus Array 1375328_at - -
Affymetrix Rat Clariom S Assay PSR1000001751.rn.1 TC1000000192.rn.1 RTA TC
Rat Expression Set 230 1375328_at - -
Rat Expression Set 230 1375328_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10940587 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21012033 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline