This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01464132_g1
See other ATP5G1 GE Assays ›
Gene Symbol:
Atp5g1
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C1 (subunit 9)
Gene Aliases:
-
Chromosome Location:
Chr.10: 83895941 - 83898640 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01464132_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5g1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006247189.1 XP_006247251.1 1 - 2 185 83

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_006247189.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP synthesis coupled proton transport
ATP hydrolysis coupled proton transport
response to ethanol
ATP metabolic process
molecular_function
lipid binding
hydrogen ion transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0