Assay Details
| Gene Symbol: Thra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017960.1 | NP_001017960.1 | 3 - 4 | 560 | 95 | |
| NM_031134.2 | NP_112396.2 | 2 - 3 | 428 | 95 | ||
| GenBank mRNA | CB580585.1 | - | 2 - 3 | 360 | 95 | |
| M18028.1 | - | 3 - 4 | 450 | 95 | ||
| M31174.1 | - | 642 | 95 | |||
| X07409.1 | - | 1 - 2 | 89 | 95 | ||
| X12744.1 | - | 2 - 3 | 92 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the X07409.1 transcripts.
Set Membership: |
|

Thra
Genomic Map