This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01464214_m1
See other RBFOX3 GE Assays ›
Gene Symbol:
Rbfox3
Gene Name:
RNA binding protein, fox-1 homolog (C. elegans) 3
Gene Aliases:
Hrnbp3, Neun, RGD1560070
Chromosome Location:
Chr.10: 107515821 - 107541447 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01464214_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbfox3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001134498.2 NP_001127970.1 4 - 5 511 82
XM_008768343.1 XP_008766565.1 4 - 5 2491 82
XM_008768344.1 XP_008766566.1 4 - 5 2490 82
XM_008768345.1 XP_008766567.1 4 - 5 2489 82
XM_008768346.1 XP_008766568.1 4 - 5 2488 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000004213.rn.1

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
RNA splicing
nucleotide binding
DNA binding
RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000038531.rn.1 TC1000004213.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0