This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01468296_g1
See other ESD GE Assays ›
Gene Symbol:
Esd
Gene Name:
esterase D
Gene Aliases:
-
Chromosome Location:
Chr.15: 56756848 - 56776348 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01468296_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Esd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106051.3 NP_001099521.1 5 - 6 435 146
NM_001270865.1 NP_001257794.1 5 - 6 517 146
NM_001270866.1 NP_001257795.1 5 - 6 435 146
GenBank mRNA BC158790.1 - 5 - 6 426 7308742 146
FQ213565.1 - 5 - 6 435 146
FQ216373.1 - 5 - 6 519 146
FQ218750.1 - 5 - 6 408 146
FQ225558.1 - 2 - 3 195 146
FQ229329.1 - 5 - 6 409 146
FQ234562.1 - 2 - 3 195 146

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
formaldehyde catabolic process
molecular_function
hydrolase activity, acting on ester bonds
S-formylglutathione hydrolase activity
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1500005711.rn.1 TC1500000835.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0