This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01468300_g1
See other ESD GE Assays ›
Gene Symbol:
Esd
Gene Name:
esterase D
Gene Aliases:
-
Chromosome Location:
Chr.15: 56756848 - 56776348 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01468300_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Esd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106051.3 NP_001099521.1 7 - 8 655 110
NM_001270865.1 NP_001257794.1 7 - 8 737 110
NM_001270866.1 NP_001257795.1 7 - 8 655 110
GenBank mRNA BC158790.1 - 7 - 8 646 7308742 110
FQ211025.1 - 1 - 2 97 110
FQ213565.1 - 7 - 8 655 110
FQ225558.1 - 4 - 5 415 110
FQ229329.1 - 7 - 8 629 110
FQ234562.1 - 4 - 5 415 110

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
formaldehyde catabolic process
molecular_function
hydrolase activity, acting on ester bonds
S-formylglutathione hydrolase activity
carboxylic ester hydrolase activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10301 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 198327 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 43372 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 391343 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 930732 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 33830 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 192278 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 327365 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 602322 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 305295 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 628653 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 762323 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 686830 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 659199 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 803261 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 915967 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 168983 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 58456 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10301 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10301 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10301 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10301 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 89027 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10781475 - -
Affymetrix Rat Gene 1.0 ST Array 10781477 - -
Affymetrix Rat Gene 1.0 ST Array 10781478 - -
Affymetrix Rat Gene 1.0 ST Array 10781479 - -
Affymetrix Rat Gene 1.0 ST Array 10781481 - -
Affymetrix Rat Gene 1.0 ST Array 10781476 - -
Affymetrix Rat Gene 1.0 ST Array 10781474 - -
Affymetrix Rat Gene 1.0 ST Array 10781480 - -
Affymetrix Rat Gene 1.1 ST Array 10781473 - -
Rat Genome 230 2.0 Array 1398903_at - -
Rat Genome 230 2.0 Array 1398903_PM_at - -
Affymetrix HT Rat Focus Array 1398903_at - -
Affymetrix HT Rat Focus Array 1398903_PM_at - -
Affymetrix Rat Clariom S Assay PSR1500005714.rn.1 TC1500000835.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500005716.rn.1 TC1500000835.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500005717.rn.1 TC1500000835.rn.1 RTA TC
Rat Expression Set 230 1398903_PM_at - -
Rat Expression Set 230 1398903_at - -
Affymetrix Rat Gene 1.0 ST Array 10781473 - -
Rat Genome U34 Set rc_AA899045_g_at - -
Rat Genome U34 Set rc_AA899045_at - -
Rat Genome U34 Set rc_AI233204_at - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1371986 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7r99j:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0