This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01468700_m1
See other SLC7A15 GE Assays ›
Gene Symbol:
Slc7a15
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 15
Gene Aliases:
RGD1560726
Chromosome Location:
Chr.6: 33728107 - 33739115 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01468700_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106714.1 NP_001100184.1 1 - 2 781 143

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0600002111.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
amino acid transmembrane transporter activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600015459.rn.1 TC0600002111.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600015461.rn.1 TC0600002111.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-97q5c:80/100.66.78.89:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0