This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01469765_g1
See other TWISTNB GE Assays ›
Gene Symbol:
Twistnb
Gene Name:
TWIST neighbor
Gene Aliases:
-
Chromosome Location:
Chr.6: 52751106 - 52768574 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01469765_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Twistnb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108707.1 NP_001102177.1 3 - 4 711 170
GenBank mRNA FQ230315.1 - 3 - 4 626 170

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0600000600.rn.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase I promoter
biological_process
RNA polymerase I activity
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1029107 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P945846 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 583280 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 903592 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 275467 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 841067 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 282744 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 528390 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 514138 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 86274 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 532958 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 793832 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 105759 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 457371 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 69870 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 138738 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1029107 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P945846 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P945846 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1029107 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P945846 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1029107 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1029107 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P915530 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 16487 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10884210 - -
Affymetrix Rat Gene 1.0 ST Array 10884207 - -
Affymetrix Rat Gene 1.0 ST Array 10884209 - -
Affymetrix Rat Gene 1.0 ST Array 10884208 - -
Affymetrix Rat Gene 1.1 ST Array 10884205 - -
Rat Genome 230 2.0 Array 1385342_at - -
Rat Genome 230 2.0 Array 1385342_PM_at - -
Rat Genome 230 2.0 Array 1395500_PM_at - -
Rat Genome 230 2.0 Array 1377842_PM_at - -
Rat Genome 230 2.0 Array 1395500_at - -
Rat Genome 230 2.0 Array 1377842_at - -
Affymetrix HT Rat Focus Array 1377842_PM_at - -
Affymetrix HT Rat Focus Array 1377842_at - -
Affymetrix Rat Clariom S Assay PSR0600004301.rn.1 TC0600000600.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600004302.rn.1 TC0600000600.rn.1 RTA TC
Rat Expression Set 230 1395500_at - -
Rat Expression Set 230 1385342_PM_at - -
Rat Expression Set 230 1395500_PM_at - -
Rat Expression Set 230 1377842_at - -
Rat Expression Set 230 1385342_at - -
Rat Expression Set 230 1377842_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10884205 - -
Rat Genome U34 Set rc_AA998932_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21947586 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1360999 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0