This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01470053_m1
See other BRE GE Assays ›
Gene Symbol:
Bre
Gene Name:
brain and reproductive organ-expressed (TNFRSF1A modulator)
Gene Aliases:
-
Chromosome Location:
Chr.6: 25666654 - 26051267 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01470053_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bre
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199270.1 NP_954891.1 7 - 8 766 156
XM_006239802.2 XP_006239864.1 8 - 9 875 156
GenBank mRNA BC061573.1 - 7 - 8 766 6887072 156

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0600003494.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bre

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair
apoptotic process
cellular response to DNA damage stimulus
response to ionizing radiation
chromatin modification
positive regulation of DNA repair
tumor necrosis factor receptor binding
polyubiquitin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600024632.rn.1 TC0600003494.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0