This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01473605_m1
See other ATP5S GE Assays ›
Gene Symbol:
Atp5s
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex, subunit s (factor B)
Gene Aliases:
-
Chromosome Location:
Chr.6: 92057786 - 92076022 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01473605_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5s
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007749.1 NP_001007750.1 1 - 2 308 140
GenBank mRNA BC083655.1 - 1 - 2 308 7135201 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0600000904.rn.1

Additional Information:

This assay is in the 5'UTR region of the BC083655.1,NM_001007749.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5s

Panther Classification:

Molecular Function -

ATP synthase primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
proton transport
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600005983.rn.1 TC0600000904.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600005982.rn.1 TC0600000904.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600005985.rn.1 TC0600000904.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600005981.rn.1 TC0600000904.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0