Assay Details
| Gene Symbol: Sdr16c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106634.1 | NP_001100104.1 | 2 - 3 | 682 | 92 | |
| XM_006237836.2 | XP_006237898.1 | 3 - 4 | 569 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sdr16c5
Genomic Map