This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01474469_m1
See other GLYATL2 GE Assays ›
Gene Symbol:
Glyatl2
Gene Name:
glycine-N-acyltransferase-like 2
Gene Aliases:
HP33, Keg1
Chromosome Location:
Chr.1: 229519586 - 229552573 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01474469_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glyatl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134330.2 NP_599157.2 4 - 5 349 91
GenBank mRNA AB019693.1 - 415 91
BC078701.1 - 4 - 5 349 7130696 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100003265.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Glyatl2

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
gamma-tubulin binding
glycine N-acyltransferase activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P118012 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 916371 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 866886 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 161463 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 286451 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 539146 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 111204 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 384792 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 945055 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 157508 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 160798 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 674974 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 411421 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 842386 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 58165 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 278590 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 743846 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 886447 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 486234 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 825958 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 559932 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 817350 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 76507 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 98854 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 57141 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 20074 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 4962 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118012 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118012 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P118012 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P517738 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 701606928 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 43321 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 62441 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 98577 - -
Agilent Rat cDNA Microarray (G4105A) 701606928 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10714134 - -
Affymetrix Rat Gene 1.0 ST Array 10714130 - -
Affymetrix Rat Gene 1.0 ST Array 10714133 - -
Affymetrix Rat Gene 1.0 ST Array 10714129 - -
Affymetrix Rat Gene 1.0 ST Array 10714132 - -
Affymetrix Rat Gene 1.0 ST Array 10714131 - -
Affymetrix Rat Gene 1.0 ST Array 10714135 - -
Affymetrix Rat Gene 1.1 ST Array 10714128 - -
Rat Genome 230 2.0 Array 1370592_at - -
Rat Genome 230 2.0 Array 1370592_PM_at - -
Affymetrix HT Rat Focus Array 1370592_PM_at - -
Affymetrix HT Rat Focus Array 1370592_at - -
Affymetrix Rat Clariom S Assay PSR0100026952.rn.1 TC0100003265.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100026951.rn.1 TC0100003265.rn.1 RTA TC
Rat Expression Set 230 1370592_at - -
Rat Expression Set 230 1370592_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10714128 - -
Rat Genome U34 Set AB019693_at - -
Rat Genome U34 Set rc_AA893035_s_at - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1355872 - -
RatRef-12 Expression BeadChip ILMN_62369 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0