This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01477344_m1
See other BACH1 GE Assays ›
Gene Symbol:
Bach1
Gene Name:
BTB and CNC homology 1, basic leucine zipper transcription factor 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 27364871 - 27398713 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01477344_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bach1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107113.1 NP_001100583.1 3 - 4 1725 115
XM_003751009.3 XP_003751057.2 3 - 4 1770 115
XM_006248018.2 XP_006248080.1 3 - 4 1770 115
XM_006248019.2 XP_006248081.1 3 - 4 2929 115
XM_006248020.2 XP_006248082.1 3 - 4 2786 115
XM_008768581.1 XP_008766803.1 3 - 4 3529 115

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1100000177.rn.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription involved in G1/S transition of mitotic cell cycle
regulation of transcription involved in G2/M transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of transcription from RNA polymerase II promoter in response to hypoxia
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P056623 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 82454 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 883207 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 598709 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 876027 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 24378 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 583745 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 573728 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 706294 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 430193 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 347356 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 10971 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 905216 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 951180 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 152444 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 382377 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 377459 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 1594 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 357560 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 545224 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 90648 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 73925 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 15890 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 161805 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 67354 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P056623 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P056623 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P056623 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P362714 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19327 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 53241 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10750082 - -
Affymetrix Rat Gene 1.0 ST Array 10750081 - -
Affymetrix Rat Gene 1.0 ST Array 10750084 - -
Affymetrix Rat Gene 1.0 ST Array 10750083 - -
Affymetrix Rat Gene 1.0 ST Array 10750085 - -
Affymetrix Rat Gene 1.1 ST Array 10750080 - -
Rat Genome 230 2.0 Array 1377699_at - -
Rat Genome 230 2.0 Array 1377699_PM_at - -
Affymetrix HT Rat Focus Array 1377699_at - -
Affymetrix HT Rat Focus Array 1377699_PM_at - -
Affymetrix Rat Clariom S Assay PSR1100000679.rn.1 TC1100000177.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100000674.rn.1 TC1100000177.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100000676.rn.1 TC1100000177.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100000677.rn.1 TC1100000177.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100000678.rn.1 TC1100000177.rn.1 RTA TC
Rat Expression Set 230 1377699_at - -
Rat Expression Set 230 1377699_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10750080 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22133702 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1371410 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0