This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01478029_m1
See other MCEE GE Assays ›
Gene Symbol:
Mcee
Gene Name:
methylmalonyl CoA epimerase
Gene Aliases:
-
Chromosome Location:
Chr.1: 125229487 - 125252692 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01478029_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mcee
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106341.1 NP_001099811.1 1 - 2 177 119
XM_008759512.1 XP_008757734.1 1 - 2 125 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100001670.rn.1

Additional Information:

This assay is in the 5'UTR region of the NM_001106341.1,XM_008759512.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

epimerase/racemase

Gene Ontology Categories:

Function(s) Process(es)

L-methylmalonyl-CoA metabolic process
methylmalonyl-CoA epimerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100013420.rn.1 TC0100001670.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100013425.rn.1 TC0100001670.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100013424.rn.1 TC0100001670.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100013419.rn.1 TC0100001670.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100013418.rn.1 TC0100001670.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0