This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01478297_m1
See other SV2B GE Assays ›
Gene Symbol:
Sv2b
Gene Name:
synaptic vesicle glycoprotein 2b
Gene Aliases:
-
Chromosome Location:
Chr.1: 136664201 - 136842679 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01478297_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sv2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_057207.3 NP_476555.1 11 - 12 2154 85
XM_008759492.1 XP_008757714.1 11 - 12 2873 85
XM_008759493.1 XP_008757715.1 11 - 12 3251 85
XM_008759494.1 XP_008757716.1 11 - 12 3773 85
XM_008759495.1 XP_008757717.1 11 - 12 2869 85
GenBank mRNA L10362.1 - 11 - 12 2154 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100005796.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sv2b

Gene Ontology Categories:

Function(s) Process(es)

neurotransmitter uptake
transmembrane transport
transmembrane transporter activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P328121 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 83669 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 336501 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 760589 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 225779 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 216117 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 312351 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 522647 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 219828 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 415591 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 71504 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 473179 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 614932 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 919647 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 377748 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 666672 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 880887 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 353166 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 452883 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 199670 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 422817 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 687692 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 946700 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 179815 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 657778 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 927807 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 728105 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 773991 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 856992 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 637311 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 773246 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 564277 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 850540 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 285002 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 420627 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 928305 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 377703 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 475728 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 841558 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 652578 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 291732 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 607271 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 800246 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 674940 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 793211 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 439676 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 174065 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 20018 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 637262 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 570166 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 557478 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 702417 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 888179 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 415449 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 972381 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 587901 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 30983 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 599084 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 380877 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 776995 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 379136 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 396027 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 390826 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 225792 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 426681 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 105236 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 561499 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 637313 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 567327 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 433917 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 230377 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 199755 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 65284 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 929606 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 549648 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 461476 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 245950 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 910494 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 279532 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 772241 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 799634 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 843005 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 608078 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 892638 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 250462 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 253761 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 815865 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 568273 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 807826 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 520651 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 36930 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 538275 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 593568 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 509484 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 704348 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 704532 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 348189 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 528944 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 513859 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 530727 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 61299 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 357558 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 463236 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 54837 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 217643 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 623005 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 835885 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 318046 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 965611 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 740371 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 809502 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 656536 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 174178 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 321905 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 612069 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 139629 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 489536 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 502149 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 853063 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 494266 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 360781 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 432187 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 732183 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 790848 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 216991 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 338231 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 684437 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 463533 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 267210 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 333754 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 178078 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 96749 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 103311 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 174012 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 109368 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 131047 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 112068 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 61134 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 171111 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 7814 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 160069 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 47262 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 49866 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 46730 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 8908 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 7370 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 30974 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 29933 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 59782 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 10331 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 126681 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 110323 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 133171 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 137238 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 31557 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 116834 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 74893 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 86393 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P328121 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P328121 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P328121 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P242712 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P328121 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P310714 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 30151 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 44203 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 67698 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 34175 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 951 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 31808 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 83156 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 85996 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 99491 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 90485 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10722838 - -
Affymetrix Rat Gene 1.0 ST Array 10722837 - -
Affymetrix Rat Gene 1.0 ST Array 10722836 - -
Affymetrix Rat Gene 1.0 ST Array 10722842 - -
Affymetrix Rat Gene 1.0 ST Array 10722835 - -
Affymetrix Rat Gene 1.0 ST Array 10722840 - -
Affymetrix Rat Gene 1.0 ST Array 10722833 - -
Affymetrix Rat Gene 1.0 ST Array 10722834 - -
Affymetrix Rat Gene 1.0 ST Array 10722841 - -
Affymetrix Rat Gene 1.0 ST Array 10722843 - -
Affymetrix Rat Gene 1.0 ST Array 10722844 - -
Affymetrix Rat Gene 1.0 ST Array 10722839 - -
Affymetrix Rat Gene 1.0 ST Array 10722831 - -
Affymetrix Rat Gene 1.0 ST Array 10722832 - -
Affymetrix Rat Gene 1.1 ST Array 10722830 - -
Rat Genome 230 2.0 Array 1369628_PM_at - -
Rat Genome 230 2.0 Array 1369628_at - -
Rat Genome 230 2.0 Array 1369627_at - -
Rat Genome 230 2.0 Array 1375142_at - -
Rat Genome 230 2.0 Array 1369627_PM_at - -
Rat Genome 230 2.0 Array 1375142_PM_at - -
Affymetrix HT Rat Focus Array 1369627_at - -
Affymetrix HT Rat Focus Array 1375142_at - -
Affymetrix HT Rat Focus Array 1375142_PM_at - -
Affymetrix HT Rat Focus Array 1369628_at - -
Affymetrix HT Rat Focus Array 1369627_PM_at - -
Affymetrix HT Rat Focus Array 1369628_PM_at - -
Affymetrix Rat Clariom S Assay PSR0100046249.rn.1 TC0100005796.rn.1 RTA TC
Rat Expression Set 230 1369628_at - -
Rat Expression Set 230 1375142_PM_at - -
Rat Expression Set 230 1369627_at - -
Rat Expression Set 230 1375142_at - -
Rat Expression Set 230 1369628_PM_at - -
Rat Expression Set 230 1369627_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10722830 - -
Rat Genome U34 Set rc_AI639484_at - -
Rat Genome U34 Set L10362_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22136159 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1364550 - -
RatRef-12 Expression BeadChip ILMN_64138 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0