This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01479169_g1
See other NUDT22 GE Assays ›
Gene Symbol:
Nudt22
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 22
Gene Aliases:
-
Chromosome Location:
Chr.1: 222247500 - 222250983 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01479169_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt22
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199090.2 NP_954521.1 1 - 2 98 70
GenBank mRNA BC060593.1 - 1 - 2 99 6887766 70
FQ212766.1 - 1 - 2 45 70
FQ219356.1 - 1 - 2 49 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100007185.rn.1

Additional Information:

This assay is in the 5'UTR region of the BC060593.1,NM_199090.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nudt22

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100057842.rn.1 TC0100007185.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100057840.rn.1 TC0100007185.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100057838.rn.1 TC0100007185.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0