This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01479928_m1
See other IFIT3 GE Assays ›
Gene Symbol:
Ifit3
Gene Name:
interferon-induced protein with tetratricopeptide repeats 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 252906234 - 252911377 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01479928_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ifit3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007694.1 NP_001007695.1 1 - 2 157 95
GenBank mRNA BC079079.1 - 1 - 2 157 7122607 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100003517.rn.1

Additional Information:

This assay is in the 5'UTR region of the BC079079.1,NM_001007694.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ifit3

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cell proliferation
response to virus
cellular response to interferon-alpha
cellular response to interferon-beta
negative regulation of apoptotic process
identical protein binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P082067 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 63406 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 303906 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 656495 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 627932 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082067 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082067 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P082067 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P283726 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 19798 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10714906 - -
Affymetrix Rat Gene 1.0 ST Array 10714905 - -
Affymetrix Rat Gene 1.0 ST Array 10714904 - -
Affymetrix Rat Gene 1.1 ST Array 10714903 - -
Rat Genome 230 2.0 Array 1376908_PM_at - -
Rat Genome 230 2.0 Array 1376908_at - -
Affymetrix HT Rat Focus Array 1376908_PM_at - -
Affymetrix HT Rat Focus Array 1376908_at - -
Affymetrix Rat Clariom S Assay PSR0100028487.rn.1 TC0100003517.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100028486.rn.1 TC0100003517.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100028485.rn.1 TC0100003517.rn.1 RTA TC
Rat Expression Set 230 1376908_at - -
Rat Expression Set 230 1376908_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10714903 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21705862 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1367152 - -
RatRef-12 Expression BeadChip ILMN_52894 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0