This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01481369_gH
See other UNC119B GE Assays ›
Gene Symbol:
Unc119b
Gene Name:
unc-119 homolog B (C. elegans)
Gene Aliases:
RGD1309359
Chromosome Location:
Chr.12: 47239663 - 47252286 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01481369_gH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Unc119b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001105934.1 NP_001099404.1 3 - 4 533 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1200000786.rn.1

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

lipoprotein transport
cilium morphogenesis
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1200006908.rn.1 TC1200000786.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200006912.rn.1 TC1200000786.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200006909.rn.1 TC1200000786.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200006910.rn.1 TC1200000786.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200006911.rn.1 TC1200000786.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0