This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01481376_m1
See other EPO GE Assays ›
Gene Symbol:
Epo
Gene Name:
erythropoietin
Gene Aliases:
-
Chromosome Location:
Chr.12: 22274828 - 22278268 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01481376_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Epo
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017001.1 NP_058697.1 4 - 5 441 64
XM_006249101.2 XP_006249163.1 3 - 4 583 64
GenBank mRNA D10763.1 - 4 - 5 441 64
L10608.1 - 3 - 4 410 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1200000374.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Epo

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
regulation of transcription from RNA polymerase II promoter
apoptotic process
embryo implantation
aging
response to nutrient
positive regulation of cell proliferation
response to salt stress
negative regulation of calcium ion transport into cytosol
peptidyl-serine phosphorylation
erythrocyte differentiation
response to lipopolysaccharide
negative regulation of myeloid cell apoptotic process
response to vitamin A
response to testosterone
positive regulation of tyrosine phosphorylation of Stat5 protein
hemoglobin biosynthetic process
negative regulation of apoptotic process
erythrocyte maturation
response to estrogen
positive regulation of neuron differentiation
positive regulation of DNA replication
positive regulation of protein kinase activity
positive regulation of transcription, DNA-templated
positive regulation of Ras protein signal transduction
response to axon injury
response to electrical stimulus
response to hyperoxia
response to interleukin-1
cellular hyperosmotic response
negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress
negative regulation of erythrocyte apoptotic process
negative regulation of cation channel activity
erythropoietin receptor binding
hormone activity
protein kinase activator activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P004043 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 84081 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 458750 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004043 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P004043 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P004043 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P320709 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 57352 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10760950 - -
Affymetrix Rat Gene 1.0 ST Array 10760951 - -
Affymetrix Rat Gene 1.0 ST Array 10760948 - -
Affymetrix Rat Gene 1.0 ST Array 10760949 - -
Affymetrix Rat Gene 1.0 ST Array 10760947 - -
Affymetrix Rat Gene 1.1 ST Array 10760946 - -
Rat Genome 230 2.0 Array 1387308_PM_at - -
Rat Genome 230 2.0 Array 1387308_at - -
Affymetrix HT Rat Focus Array 1387308_PM_at - -
Affymetrix HT Rat Focus Array 1387308_at - -
Affymetrix Rat Clariom S Assay PSR1200003162.rn.1 TC1200000374.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200003157.rn.1 TC1200000374.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200003161.rn.1 TC1200000374.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200003159.rn.1 TC1200000374.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200003160.rn.1 TC1200000374.rn.1 RTA TC
Rat Expression Set 230 1387308_PM_at - -
Rat Expression Set 230 1387308_at - -
Affymetrix Rat Gene 1.0 ST Array 10760946 - -
Rat Genome U34 Set D10763_at - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1351138 - -
RatRef-12 Expression BeadChip ILMN_63491 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline