This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01482159_m1
See other GSR GE Assays ›
Gene Symbol:
Gsr
Gene Name:
glutathione reductase
Gene Aliases:
-
Chromosome Location:
Chr.16: 62197617 - 62239987 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01482159_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gsr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053906.2 NP_446358.2 4 - 5 207 64
GenBank mRNA FM035154.1 - 4 - 5 207 64
U73174.1 - 334 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1600001797.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gsr

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
spermatogenesis
response to vitamin A
response to nicotine
cell redox homeostasis
response to ethanol
oxidation-reduction process
glutathione-disulfide reductase activity
protein homodimerization activity
glutathione binding
flavin adenine dinucleotide binding
NADP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1600013226.rn.1 TC1600001797.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600013227.rn.1 TC1600001797.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600013228.rn.1 TC1600001797.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600013229.rn.1 TC1600001797.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600013230.rn.1 TC1600001797.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline