This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01482587_m1
See other GTF2E2 GE Assays ›
Gene Symbol:
Gtf2e2
Gene Name:
general transcription factor IIE, polypeptide 2, beta
Gene Aliases:
-
Chromosome Location:
Chr.16: 62113846 - 62164333 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01482587_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2e2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107318.2 NP_001100788.2 3 - 4 303 119
XM_006253230.2 XP_006253292.1 3 - 4 368 119
XM_006253231.2 XP_006253293.1 3 - 4 368 119
GenBank mRNA CF110694.1 - 3 - 4 319 119
FQ225698.1 - 2 - 3 279 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1600001795.rn.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription initiation from RNA polymerase II promoter
biological_process
molecular_function
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1600013207.rn.1 TC1600001795.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600013208.rn.1 TC1600001795.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600013204.rn.1 TC1600001795.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600013205.rn.1 TC1600001795.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1600013206.rn.1 TC1600001795.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline