This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01483883_m1
See other METTL13 GE Assays ›
Gene Symbol:
Mettl13
Gene Name:
methyltransferase like 13
Gene Aliases:
RGD1311526
Chromosome Location:
Chr.13: 80427595 - 80441352 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01483883_m1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mettl13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001134501.1 NP_001127973.1 1 - 2 416 70
XM_006250152.2 XP_006250214.1 2 - 3 362 70
XM_008769622.1 XP_008767844.1 1 - 2 464 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1300001770.rn.1

Set Membership:

Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1300012468.rn.1 TC1300001770.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300012469.rn.1 TC1300001770.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300012471.rn.1 TC1300001770.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300012470.rn.1 TC1300001770.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fbxsq:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline