Assay Details
| Gene Symbol: Mettl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134501.1 | NP_001127973.1 | 5 - 6 | 1766 | 66 | |
| XM_006250152.2 | XP_006250214.1 | 6 - 7 | 1694 | 66 | ||
| XM_008769622.1 | XP_008767844.1 | 5 - 6 | 1796 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mettl13
Genomic Map