This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01484730_m1
See other XIRP2 GE Assays ›
Gene Symbol:
Xirp2
Gene Name:
xin actin-binding repeat containing 2
Gene Aliases:
Cmya3, LACS
Chromosome Location:
Chr.3: 53563245 - 53658113 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01484730_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Xirp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201989.1 NP_973718.1 1 - 2 134 67
GenBank mRNA AF466694.1 - 1 - 2 134 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF466694.1, NM_201989.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Xirp2

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

ventricular septum development
heart development
actin cytoskeleton organization
cell-cell junction organization
cardiac muscle tissue morphogenesis
actin binding
alpha-actinin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0300005370.rn.1 TC0300000721.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300005371.rn.1 TC0300000721.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0