This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01484735_m1
See other XIRP2 GE Assays ›
Gene Symbol:
Xirp2
Gene Name:
xin actin-binding repeat containing 2
Gene Aliases:
Cmya3, LACS
Chromosome Location:
Chr.3: 53563245 - 53658113 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01484735_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Xirp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201989.1 NP_973718.1 6 - 7 724 66
GenBank mRNA AF466694.1 - 6 - 7 724 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300000721.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Xirp2

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

ventricular septum development
heart development
actin cytoskeleton organization
cell-cell junction organization
cardiac muscle tissue morphogenesis
actin binding
alpha-actinin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0300005377.rn.1 TC0300000721.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300005378.rn.1 TC0300000721.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300005379.rn.1 TC0300000721.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300005380.rn.1 TC0300000721.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300005376.rn.1 TC0300000721.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline