This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01486272_g1
See other EIF3J GE Assays ›
Gene Symbol:
Eif3j
Gene Name:
eukaryotic translation initiation factor 3, subunit J
Gene Aliases:
Eif3s1
Chromosome Location:
Chr.3: 113976687 - 113998925 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01486272_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif3j
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077670.1 NP_001071138.1 7 - 8 745 104
GenBank mRNA BC127509.1 - 7 - 8 745 7454886 104
FQ226401.1 - 7 - 8 694 105

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
cytoplasmic translation
regulation of translational initiation
translation initiation factor activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P000245 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P309398 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 272869 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 657550 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 682532 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 771488 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 136774 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 756522 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 921871 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 672648 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 174632 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 925705 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 2005 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 282152 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 907175 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 939592 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 495944 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 163655 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 337454 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 128322 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 6196 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 138665 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000245 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P309398 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000245 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P309398 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P000245 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P309398 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P309398 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 45262 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10839223 - -
Affymetrix Rat Gene 1.0 ST Array 10788085 - -
Affymetrix Rat Gene 1.0 ST Array 10839224 - -
Affymetrix Rat Gene 1.0 ST Array 10839227 - -
Affymetrix Rat Gene 1.0 ST Array 10839228 - -
Affymetrix Rat Gene 1.0 ST Array 10839226 - -
Affymetrix Rat Gene 1.0 ST Array 10839225 - -
Affymetrix Rat Gene 1.1 ST Array 10839222 - -
Affymetrix Rat Gene 1.1 ST Array 10788079 - -
Rat Genome 230 2.0 Array 1388424_at - -
Rat Genome 230 2.0 Array 1388424_PM_at - -
Rat Genome 230 2.0 Array 1395199_at - -
Rat Genome 230 2.0 Array 1395199_PM_at - -
Affymetrix HT Rat Focus Array 1395199_at - -
Affymetrix HT Rat Focus Array 1388424_PM_at - -
Affymetrix HT Rat Focus Array 1388424_at - -
Affymetrix HT Rat Focus Array 1395199_PM_at - -
Affymetrix Rat Clariom S Assay PSR0300011011.rn.1 TC0300001580.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300011012.rn.1 TC0300001580.rn.1 RTA TC
Rat Expression Set 230 1388424_at - -
Rat Expression Set 230 1388424_PM_at - -
Rat Expression Set 230 1395199_PM_at - -
Rat Expression Set 230 1395199_at - -
Affymetrix Rat Gene 1.0 ST Array 10839222 - -
Rat Genome U34 Set rc_AI009710_at - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1360129 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0