This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01486617_m1
See other ICAM5 GE Assays ›
Gene Symbol:
Icam5
Gene Name:
intercellular adhesion molecule 5, telencephalin
Gene Aliases:
-
Chromosome Location:
Chr.8: 22050222 - 22057209 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01486617_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Icam5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001172079.1 NP_001165550.1 10 - 11 2718 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0800000216.rn.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

phagocytosis
cell adhesion
single organismal cell-cell adhesion
integrin binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P063229 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063229 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P063229 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P063229 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P394134 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1024907 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.1 ST Array 10908328 - -
Rat Genome 230 2.0 Array 1393710_at - -
Rat Genome 230 2.0 Array 1378712_PM_at - -
Rat Genome 230 2.0 Array 1393710_PM_at - -
Rat Genome 230 2.0 Array 1378712_at - -
Affymetrix HT Rat Focus Array 1393710_PM_at - -
Affymetrix HT Rat Focus Array 1378712_at - -
Affymetrix HT Rat Focus Array 1378712_PM_at - -
Affymetrix HT Rat Focus Array 1393710_at - -
Affymetrix Rat Clariom S Assay PSR0800001153.rn.1 TC0800000216.rn.1 RTA TC
Rat Expression Set 230 1393710_at - -
Rat Expression Set 230 1378712_at - -
Rat Expression Set 230 1378712_PM_at - -
Rat Expression Set 230 1393710_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21148195 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1352283 - -
RatRef-12 Expression BeadChip ILMN_69263 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0