This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01487276_m1
See other PRTFDC1 GE Assays ›
Gene Symbol:
Prtfdc1
Gene Name:
phosphoribosyl transferase domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.17: 87926242 - 88037051 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01487276_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prtfdc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106127.1 NP_001099597.1 1 - 2 85 82
XM_008771860.1 XP_008770082.1 1 - 2 96 82
GenBank mRNA BC166887.1 - 1 - 2 82 6890546 82
BC166923.1 - 1 - 2 82 6919422 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1700002346.rn.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

purine ribonucleoside salvage
adenine salvage
guanine salvage
GMP salvage
IMP salvage
GMP catabolic process
hypoxanthine metabolic process
magnesium ion binding
hypoxanthine phosphoribosyltransferase activity
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1700015684.rn.1 TC1700002346.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700015686.rn.1 TC1700002346.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700015687.rn.1 TC1700002346.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0