This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01487285_g1
See other ATP5C1 GE Assays ›
Gene Symbol:
Atp5c1
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1
Gene Aliases:
-
Chromosome Location:
Chr.17: 72209320 - 72231566 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01487285_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5c1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053825.2 NP_446277.1 1 - 2 93 55
XM_006254229.2 XP_006254291.1 1 - 2 147 55
XM_006254230.2 XP_006254292.1 1 - 2 148 55
GenBank mRNA BC059158.1 - 1 - 2 97 6920493 55
FQ212577.1 - 1 - 2 81 55
FQ212651.1 - 1 - 2 91 55
FQ212813.1 - 1 - 2 91 55
FQ213008.1 - 1 - 2 91 55
FQ213785.1 - 1 - 2 93 55
FQ213883.1 - 1 - 2 87 55
FQ214745.1 - 1 - 2 71 55
FQ214863.1 - 1 - 2 147 55
FQ214998.1 - 1 - 2 91 55
FQ215205.1 - 1 - 2 91 55
FQ215286.1 - 1 - 2 66 55
FQ215351.1 - 1 - 2 91 55
FQ215426.1 - 1 - 2 90 55
FQ215847.1 - 1 - 2 91 55
FQ216198.1 - 1 - 2 87 55
FQ216285.1 - 1 - 2 81 55
FQ216490.1 - 1 - 2 91 55
FQ216539.1 - 1 - 2 75 55
FQ216647.1 - 1 - 2 93 55
FQ216736.1 - 1 - 2 91 55
FQ218995.1 - 1 - 2 93 55
FQ219484.1 - 1 - 2 91 55
FQ219563.1 - 1 - 2 91 55
FQ219689.1 - 1 - 2 93 55
FQ220076.1 - 1 - 2 91 55
FQ220217.1 - 1 - 2 93 55
FQ220355.1 - 1 - 2 91 55
FQ224939.1 - 1 - 2 91 55
FQ229987.1 - 1 - 2 114 55
FQ234981.1 - 1 - 2 91 55

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5c1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

oxidative phosphorylation
ATP synthesis coupled proton transport
ATP metabolic process
ATPase activity
poly(A) RNA binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1700006529.rn.1 TC1700000945.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700006523.rn.1 TC1700000945.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0