This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01487516_gH
See other AKR1C2 GE Assays ›
Gene Symbol:
Akr1c2
Gene Name:
aldo-keto reductase family 1, member C2
Gene Aliases:
Akr1c21
Chromosome Location:
Chr.17: 69388337 - 69435160 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01487516_gH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Akr1c2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013057.1 NP_001013075.1 5 - 6 640 117
XM_006254183.2 XP_006254245.1 5 - 6 652 117
XM_006254184.2 XP_006254246.1 5 - 6 631 117
GenBank mRNA BC078957.1 - 5 - 6 640 7110806 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1700002053.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Akr1c2

Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

prostaglandin metabolic process
G-protein coupled receptor signaling pathway
digestion
steroid metabolic process
positive regulation of cell proliferation
epithelial cell differentiation
progesterone metabolic process
daunorubicin metabolic process
doxorubicin metabolic process
positive regulation of protein kinase B signaling
oxidation-reduction process
cellular response to jasmonic acid stimulus
cellular response to prostaglandin D stimulus
alditol:NADP+ 1-oxidoreductase activity
oxidoreductase activity
oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
phenanthrene 9,10-monooxygenase activity
carboxylic acid binding
bile acid binding
steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
androsterone dehydrogenase activity
ketosteroid monooxygenase activity
trans-1,2-dihydrobenzene-1,2-diol dehydrogenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1700014102.rn.1 TC1700002053.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0