This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01487534_m1
See other UCMA GE Assays ›
Gene Symbol:
Ucma
Gene Name:
upper zone of growth plate and cartilage matrix associated
Gene Aliases:
GRP, RGD1308977
Chromosome Location:
Chr.17: 77252099 - 77262296 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01487534_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ucma
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106121.1 NP_001099591.1 1 - 2 184 56
XM_006254265.1 XP_006254327.1 3 - 4 824 56
GenBank mRNA EU022754.1 - 3 - 4 259 56

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1700002210.rn.1

Set Membership:

Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s)

negative regulation of osteoblast differentiation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1700014744.rn.1 TC1700002210.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700014741.rn.1 TC1700002210.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700014743.rn.1 TC1700002210.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0