This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01487697_m1
See other HACD1 GE Assays ›
Gene Symbol:
Hacd1
Gene Name:
3-hydroxyacyl-CoA dehydratase 1
Gene Aliases:
Ptpla
Chromosome Location:
Chr.17: 81173238 - 81196012 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01487697_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hacd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001101001.1 NP_001094471.1 4 - 5 454 82
XM_006254292.2 XP_006254354.1 3 - 4 1127 82
GenBank mRNA AM260199.1 - 4 - 5 454 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1700002258.rn.1

Set Membership:

Amplicon length between 71 and 85 Protein Phosphatase Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of G2/M transition of mitotic cell cycle
positive regulation of cell-substrate adhesion
myotube differentiation
sphingolipid biosynthetic process
fatty acid elongation
cellular macromolecular complex assembly
very long-chain fatty acid biosynthetic process
cementum mineralization
regulation of G1/S transition of mitotic cell cycle
3-hydroxyacyl-CoA dehydratase activity
enzyme binding
hydroxyapatite binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1700015242.rn.1 TC1700002258.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1700015244.rn.1 TC1700002258.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0