Assay Details
| Gene Symbol: Cyp4v3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135600.1 | NP_001129072.1 | 10 - 11 | 1644 | 71 | |
| GenBank mRNA | BC131846.1 | - | 10 - 11 | 1644 | 7098966 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P297252 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P355775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51286 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P355775 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P297252 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P355775 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P297252 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P297252 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P355775 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P297252 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15735 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701093492 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700606449 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701467987 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10654 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701467987 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700606449 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701093492 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10788273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788277 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788275 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788267 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788272 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788268 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788271 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788269 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788270 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10788246 | - | - |
| Rat Genome 230 2.0 Array | 1388127_at | - | - |
| Rat Genome 230 2.0 Array | 1370889_at | - | - |
| Rat Genome 230 2.0 Array | 1388127_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370889_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388127_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370889_at | - | - |
| Affymetrix HT Rat Focus Array | 1370889_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388127_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1600004767.rn.1 | TC1600000594.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600004775.rn.1 | TC1600000594.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600004770.rn.1 | TC1600000594.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600004774.rn.1 | TC1600000594.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600004766.rn.1 | TC1600000594.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600004771.rn.1 | TC1600000594.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600004772.rn.1 | TC1600000594.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600004768.rn.1 | TC1600000594.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600004769.rn.1 | TC1600000594.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600004773.rn.1 | TC1600000594.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600004776.rn.1 | TC1600000594.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370889_PM_at | - | - |
| Rat Expression Set 230 | 1388127_PM_at | - | - |
| Rat Expression Set 230 | 1388127_at | - | - |
| Rat Expression Set 230 | 1370889_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788246 | - | - |
| Rat Genome U34 Set | rc_AA946300_at | - | - |
| Rat Genome U34 Set | rc_AA962978_at | - | - |
| Rat Genome U34 Set | rc_AI071323_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353124 | - | - |

Cyp4v3
Genomic Map