This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01488573_g1
See other SDR39U1 GE Assays ›
Gene Symbol:
Sdr39u1
Gene Name:
short chain dehydrogenase/reductase family 39U, member 1
Gene Aliases:
RGD1309307
Chromosome Location:
Chr.15: 34563786 - 34567465 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01488573_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sdr39u1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108378.1 NP_001101848.1 2 - 3 262 107
XM_006251992.2 XP_006252054.1 2 - 3 194 107
XM_006251993.1 XP_006252055.1 2 - 3 766 107
XM_006251994.2 XP_006252056.1 2 - 3 333 107
XM_008770689.1 XP_008768911.1 2 - 3 328 107
GenBank mRNA BC167001.1 - 2 - 3 285 107
FQ223994.1 - 1 - 2 133 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1500001731.rn.1

Additional Information:

This assay is in the 5'UTR region of the XM_006251993.1,XM_006251994.2,XM_008770689.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

epimerase/racemase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
catalytic activity
coenzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1500011384.rn.1 TC1500001731.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500011380.rn.1 TC1500001731.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500011379.rn.1 TC1500001731.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline