This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01488576_g1
See other SDR39U1 GE Assays ›
Gene Symbol:
Sdr39u1
Gene Name:
short chain dehydrogenase/reductase family 39U, member 1
Gene Aliases:
RGD1309307
Chromosome Location:
Chr.15: 34563786 - 34567465 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01488576_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sdr39u1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108378.1 NP_001101848.1 5 - 6 617 97
XM_006251992.2 XP_006252054.1 5 - 6 549 97
XM_006251993.1 XP_006252055.1 5 - 6 1176 97
XM_006251994.2 XP_006252056.1 5 - 6 743 97
XM_008770689.1 XP_008768911.1 5 - 6 738 97
GenBank mRNA BC167001.1 - 4 - 5 915 97
FQ223994.1 - 4 - 5 488 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1500001731.rn.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

epimerase/racemase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
catalytic activity
coenzyme binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P053130 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 631630 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 929595 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 58269 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053130 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P053130 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P053130 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1004282 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10783983 - -
Affymetrix Rat Gene 1.0 ST Array 10783982 - -
Affymetrix Rat Gene 1.0 ST Array 10783987 - -
Affymetrix Rat Gene 1.0 ST Array 10783984 - -
Affymetrix Rat Gene 1.0 ST Array 10783986 - -
Affymetrix Rat Gene 1.0 ST Array 10783985 - -
Affymetrix Rat Gene 1.1 ST Array 10783981 - -
Rat Genome 230 2.0 Array 1393673_at - -
Rat Genome 230 2.0 Array 1376222_PM_at - -
Rat Genome 230 2.0 Array 1393673_PM_at - -
Rat Genome 230 2.0 Array 1376990_at - -
Rat Genome 230 2.0 Array 1376990_PM_at - -
Rat Genome 230 2.0 Array 1376222_at - -
Affymetrix HT Rat Focus Array 1393673_at - -
Affymetrix HT Rat Focus Array 1393673_PM_at - -
Affymetrix HT Rat Focus Array 1376222_at - -
Affymetrix HT Rat Focus Array 1376222_PM_at - -
Affymetrix Rat Clariom S Assay PSR1500011364.rn.1 TC1500001731.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500011358.rn.1 TC1500001731.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500011362.rn.1 TC1500001731.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500011365.rn.1 TC1500001731.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500011363.rn.1 TC1500001731.rn.1 RTA TC
Rat Expression Set 230 1376222_at - -
Rat Expression Set 230 1376222_PM_at - -
Rat Expression Set 230 1393673_PM_at - -
Rat Expression Set 230 1393673_at - -
Affymetrix Rat Gene 1.0 ST Array 10783981 - -
Rat Genome U34 Set rc_AI137925_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21390653 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1370543 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline