This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01488826_g1
See other DHRS2 GE Assays ›
Gene Symbol:
Dhrs2
Gene Name:
dehydrogenase/reductase (SDR family) member 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 33876994 - 33891564 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01488826_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dhrs2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001078405.6 XP_001078405.4 9 - 10 863 86
XM_008770715.1 XP_008768937.1 9 - 10 879 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

response to toxic substance
cellular response to oxidative stress
myeloid dendritic cell differentiation
negative regulation of apoptotic process
oxidation-reduction process
carbonyl reductase (NADPH) activity

MicroArray Probes:

Agilent ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P478125 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P478125 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P478125 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20699905 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0