This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01489483_m1
See other DIMT1 GE Assays ›
Gene Symbol:
Dimt1
Gene Name:
DIM1 dimethyladenosine transferase 1 homolog (S. cerevisiae)
Gene Aliases:
Dimt1l, RGD1311752
Chromosome Location:
Chr.2: 38120558 - 38145747 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01489483_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dimt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106408.2 NP_001099878.2 13 - 14 1224 67
XM_006231902.2 XP_006231964.1 13 - 14 1252 67
XM_006231903.2 XP_006231965.1 14 - 15 1435 67
XM_006231905.2 XP_006231967.1 12 - 13 1138 67
GenBank mRNA BC158729.1 - 13 - 14 1224 9091721 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200000329.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

RNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

rRNA methylation
rRNA (adenine-N6,N6-)-dimethyltransferase activity
poly(A) RNA binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P524471 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 734092 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 466982 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 383986 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 914130 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 349581 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 606941 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 822619 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 136084 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 23927 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 68541 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 517350 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 94218 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 85661 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 744230 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 613057 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 181053 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 825318 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 81319 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 751109 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 110682 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 2577 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 20442 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P524471 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P524471 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P524471 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P524471 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P524472 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P987655 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 74386 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10812887 - -
Affymetrix Rat Gene 1.0 ST Array 10812888 - -
Affymetrix Rat Gene 1.0 ST Array 10812891 - -
Affymetrix Rat Gene 1.0 ST Array 10812895 - -
Affymetrix Rat Gene 1.0 ST Array 10812897 - -
Affymetrix Rat Gene 1.0 ST Array 10812896 - -
Affymetrix Rat Gene 1.0 ST Array 10812898 - -
Affymetrix Rat Gene 1.0 ST Array 10812894 - -
Affymetrix Rat Gene 1.0 ST Array 10812889 - -
Affymetrix Rat Gene 1.0 ST Array 10812893 - -
Affymetrix Rat Gene 1.0 ST Array 10812892 - -
Affymetrix Rat Gene 1.0 ST Array 10812890 - -
Affymetrix Rat Gene 1.1 ST Array 10812886 - -
Rat Genome 230 2.0 Array 1392532_PM_at - -
Rat Genome 230 2.0 Array 1383156_at - -
Rat Genome 230 2.0 Array 1381248_at - -
Rat Genome 230 2.0 Array 1392532_at - -
Rat Genome 230 2.0 Array 1383156_PM_at - -
Rat Genome 230 2.0 Array 1391427_PM_at - -
Rat Genome 230 2.0 Array 1385711_PM_at - -
Rat Genome 230 2.0 Array 1391427_at - -
Rat Genome 230 2.0 Array 1385711_at - -
Rat Genome 230 2.0 Array 1381248_PM_at - -
Affymetrix HT Rat Focus Array 1385711_at - -
Affymetrix HT Rat Focus Array 1391427_PM_at - -
Affymetrix HT Rat Focus Array 1391427_at - -
Affymetrix HT Rat Focus Array 1385711_PM_at - -
Affymetrix Rat Clariom S Assay PSR0200001903.rn.1 TC0200000329.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200001905.rn.1 TC0200000329.rn.1 RTA TC
Rat Expression Set 230 1385711_PM_at - -
Rat Expression Set 230 1391427_at - -
Rat Expression Set 230 1385711_at - -
Rat Expression Set 230 1391427_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10812886 - -
Rat Genome U34 Set rc_AI236342_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21379192 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1360491 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0