This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01489652_m1
See other SSBP2 GE Assays ›
Gene Symbol:
Ssbp2
Gene Name:
single-stranded DNA binding protein 2
Gene Aliases:
RGD1562968
Chromosome Location:
Chr.2: 20506981 - 20707838 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01489652_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssbp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008760646.1 XP_008758868.1 2 - 3 305 63
XM_008760647.1 XP_008758869.1 2 - 3 305 63
XM_008760648.1 XP_008758870.1 2 - 3 304 63
XM_008760649.1 XP_008758871.1 2 - 3 304 63
XM_008760650.1 XP_008758872.1 2 - 3 305 63
XM_008760651.1 XP_008758873.1 2 - 3 304 63
XM_008760652.1 XP_008758874.1 2 - 3 304 63
XM_008760653.1 XP_008758875.1 2 - 3 303 63
GenBank mRNA FQ221531.1 - 2 - 3 204 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Process(es)

single-stranded DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-ln7zh:80/100.66.79.20:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0