This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01490252_m1
See other VAMP4 GE Assays ›
Gene Symbol:
Vamp4
Gene Name:
vesicle-associated membrane protein 4
Gene Aliases:
-
Chromosome Location:
Chr.13: 80460699 - 80483597 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01490252_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vamp4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108856.1 NP_001102326.1 2 - 3 177 112
XM_006250184.2 XP_006250246.1 2 - 3 264 112
XM_006250186.2 XP_006250248.1 2 - 3 256 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1300000628.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
exocytosis
ER to Golgi vesicle-mediated transport
vesicle fusion
SNARE complex assembly
regulation of Golgi to plasma membrane protein transport
Golgi to plasma membrane protein transport
Golgi ribbon formation
toxin transport
SNARE binding
SNAP receptor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1300004363.rn.1 TC1300000628.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300004358.rn.1 TC1300000628.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300004362.rn.1 TC1300000628.rn.1 RTA TC
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20952622 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0