This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01490495_g1
See other TMEM183A GE Assays ›
Gene Symbol:
Tmem183a
Gene Name:
transmembrane protein 183A
Gene Aliases:
RGD1309892, Tmem183
Chromosome Location:
Chr.13: 51184647 - 51201400 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01490495_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem183a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013871.1 NP_001013893.1 5 - 6 742 98
XM_006249849.2 XP_006249911.1 5 - 6 806 98
GenBank mRNA BC079375.1 - 5 - 6 742 7110239 98
FQ210515.1 - 5 - 6 775 98

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem183a

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1300009704.rn.1 TC1300001440.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300009707.rn.1 TC1300001440.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1300009703.rn.1 TC1300001440.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0