This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01492087_m1
See other PPIC GE Assays ›
Gene Symbol:
Ppic
Gene Name:
peptidylprolyl isomerase C
Gene Aliases:
-
Chromosome Location:
Chr.18: 48372039 - 48384645 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01492087_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppic
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004215.1 NP_001004215.1 4 - 5 592 93
GenBank mRNA BC078949.1 - 4 - 5 592 7109986 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1800001435.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppic

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
peptidyl-prolyl cis-trans isomerase activity
cyclosporin A binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P101346 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 856493 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 662630 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 123046 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 218566 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 367750 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 723911 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 170146 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 483865 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 406695 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 444202 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 40918 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 85278 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P101346 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P101346 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P101346 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P636232 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 44538 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 16988 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10804485 - -
Affymetrix Rat Gene 1.0 ST Array 10804481 - -
Affymetrix Rat Gene 1.0 ST Array 10804484 - -
Affymetrix Rat Gene 1.0 ST Array 10804482 - -
Affymetrix Rat Gene 1.0 ST Array 10804483 - -
Affymetrix Rat Gene 1.1 ST Array 10804480 - -
Rat Genome 230 2.0 Array 1375267_PM_at - -
Rat Genome 230 2.0 Array 1375267_at - -
Affymetrix HT Rat Focus Array 1375267_PM_at - -
Affymetrix HT Rat Focus Array 1375267_at - -
Affymetrix Rat Clariom S Assay PSR1800010162.rn.1 TC1800001435.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800010165.rn.1 TC1800001435.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1800010163.rn.1 TC1800001435.rn.1 RTA TC
Rat Expression Set 230 1375267_PM_at - -
Rat Expression Set 230 1375267_at - -
Affymetrix Rat Gene 1.0 ST Array 10804480 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21914255 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1358331 - -
RatRef-12 Expression BeadChip ILMN_55034 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0