This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01493247_m1
See other GPR18 GE Assays ›
Gene Symbol:
Gpr18
Gene Name:
G protein-coupled receptor 18
Gene Aliases:
-
Chromosome Location:
Chr.15: 108323157 - 108326907 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01493247_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001079710.1 NP_001073178.1 1 - 2 184 56
GenBank mRNA BC128782.1 - 1 - 2 184 7375738 56

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1500002433.rn.1

Additional Information:

This assay is in the 5'UTR region of the BC128782.1,NM_001079710.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P164804 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P005857 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164804 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005857 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164804 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005857 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P005857 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P164804 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P431296 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10786026 - -
Affymetrix Rat Gene 1.0 ST Array 10786024 - -
Affymetrix Rat Gene 1.0 ST Array 10786025 - -
Affymetrix Rat Gene 1.0 ST Array 10786027 - -
Affymetrix Rat Gene 1.1 ST Array 10786023 - -
Affymetrix Rat Clariom S Assay PSR1500016344.rn.1 TC1500002433.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1500016343.rn.1 TC1500002433.rn.1 RTA TC
Affymetrix Rat Gene 1.0 ST Array 10786023 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0